3-Chlor-5-methyl-4-hydroxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS12574G1 AS12574G5
ACTIAS12574G1EA 51 CHF
ACTIAS12574G1 ACTIAS12574G5
3-Chlor-5-methyl-4-hydroxybenzonitril
3-Chlor-5-methyl-4-hydroxybenzonitril
Formel: C₈H₆ClNO
MDL Number: MFCD08689583
CAS-Nummer: 173900-45-3

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR