Cilnidipin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 3-Cinnamyl 5-(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-Methoxyethyl (2E)-3-Phenyl-2-propenyl Ester, 2-Methoxyethyl (2E)-3-Phenyl-2-propenyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

461750010 461750050
ACRO461750010EA 55.6 CHF
ACRO461750010 ACRO461750050
Cilnidipin 99%
Cilnidipine
Formel: C₂₇H₂₈N₂O₇
MW: 492.53 g/mol
Melting Pt: 110 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00865853
CAS-Nummer: 132203-70-4

Order Now

Spezifikation Testergebnisse

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 108 to 112 °C
Loss on drying ≤1 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR