Florisil® 60-100 mesh Pest_Residual

Lieferant: Thermo Fisher Scientific

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

457725000
ACRO457725000EA 245 CHF
ACRO457725000
Florisil® 60-100 mesh Pest_Residual
Magnesiumsilikat
Formel: MgO₃Si
MW: 100.38 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00078249
CAS-Nummer: 1343-88-0

Order Now

Spezifikation Testergebnisse

Appearance White Powder (May contain black particles)
Silicate (as SiO2) 83.5 to 84.5 %
Sodium sulfate (Na2SO4) ≤1.0 %
Magnesium oxide (MgO) 15 to 16 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR