Magnesiumsulfat, anhydrous 99% analysis_for

Lieferant: Thermo Fisher Scientific

Synonyms: MgSO4, Magnesium sulphate anhydrous

446890010 446892500
ACRO446890010EA 301 CHF
ACRO446890010 ACRO446892500
Magnesiumsulfat, anhydrous 99% analysis_for
Magnesium sulphate
Formel: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1124 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011110
CAS-Nummer: 7487-88-9
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance White to light grey Powder
Titration Complexometric ≥98.5 %
Chloride (Cl) ≤50 ppm
Arsenic (As) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Loss on ignition ≤2 % (600°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR