(R)-2-(Trifluoromethyl)piperidine 97%

Lieferant: Thermo Fisher Scientific
Gefahr

446690010
ACRO446690010EA 171 CHF
ACRO446690010
(R)-2-(Trifluoromethyl)piperidine 97%
(R)-2-(Trifluoromethyl)piperidine
Formel: C₆H₁₀F₃N
Siedepunkt: 80 °C (200Torr)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD11226598
CAS-Nummer: 1212734-83-2
UN: 2733
ADR: 3,III

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation +10° to +20° (20°C, 589 nm) (c=4 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR