Boc-Lys(Fmoc)-OH 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: N-epsilon-FMOC-N-alpha-BOC-L-Lysine

439080050
ACRO439080050EA 156 CHF
ACRO439080050
Boc-Lys(Fmoc)-OH 99%
Boc-Lys(Fmoc)-OH
Formel: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Melting Pt: 135…138 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00062032
CAS-Nummer: 84624-27-1

Order Now

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +2.3° to +4.3° (20°C, 589 nm) (c=1 in Ethyl acetate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR