5-Cyano-3-pyridinylboronic acid 97%

Lieferant: Thermo Scientific
Achtung

430400010
ACRO430400010EA 247 CHF
ACRO430400010
5-Cyano-3-pyridinylboronic acid 97%
5-Cyano-3-pyridinylboronic acid
Formel: C6H5BN2O2
MW: 147,9300 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07374882
CAS-Nummer: 497147-93-0

Order Now

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR