Methyllinoleat 99%

Lieferant: Thermo Fisher Scientific

Synonyms: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester

428130250 428130050 428131000
ACRO428130250EA 553 CHF
ACRO428130250 ACRO428130050 ACRO428131000
Methyllinoleat 99%
Methyllinoleat
Formel: C₁₉H₃₄O₂
Siedepunkt: 212 °C (16 mmHg)
Melting Pt: –35 °C
Dichte: 0.89 g/cm³
Flash-Pt: >200 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00009534
CAS-Nummer: 112-63-0
EINECS: 203-993-0

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -85° to -95° (20°C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR