4-Nitrosalicylsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 2-Hydroxy-4-nitrobenzoic acid

425870250
ACRO425870250EA 280 CHF
ACRO425870250
4-Nitrosalicylsäure 97%
4-Nitrosalicylsäure
Formel: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 235…239 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00071505
CAS-Nummer: 619-19-2

Order Now

Spezifikation Testergebnisse

Appearance White to yellow-grey to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 230 to 239 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR