Nitroprussid-Natrium Dihydrat ACS, meetSpec_Reagent Ph.Eur.

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Sodium pentacyanonitrosylferrate(III) dihydrate, Sodium nitrosopentacyanoferrate (III) dihydrate, Sodium nitroferricyanide(III) dihydrate, Disodium pentacyanonitrosylferrate (III) dihydrate, Sodium nitroferricyanide dihydrate

424360250 424361000 424365000 424360025
ACRO424360250EA 51.8 CHF
ACRO424360250 ACRO424361000 ACRO424365000 ACRO424360025
Nitroprussid-Natrium Dihydrat ACS, meetSpec_Reagent Ph.Eur.
Nitroprussid-Natrium Dihydrat
Formel: Na₂[Fe(CN)₅NO]·2H₂O
MW: 297.95 g/mol
Dichte: 1.72 g/cm³
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149192
CAS-Nummer: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1 III
Merck Index: 13,08722

Order Now

Spezifikation Testergebnisse

Appearance Bordeaux Crystals
Titration Argentometric 99.0 to 102.0 %
Chloride (Cl) ≤0.02 %
Sulfate (SO4) Passes test (Limit about 0.01%)
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR