Eisen(III)chlorid Hexahydrat 97+% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Iron trichloride hexahydrate, Ferric chloride hexahydrate

423700050 423705000 423700025 423700100
ACRO423700050EA 24.4 CHF
ACRO423700050 ACRO423705000 ACRO423700025 ACRO423700100
Eisen(III)chlorid Hexahydrat 97+% ACS
Eisen(III)chlorid Hexahydrat
Formel: FeCl₃·6H₂O
Siedepunkt: 280…285 °C
Melting Pt: 37 °C
Dichte: 1,82 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149712
CAS-Nummer: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Yellow to orange to light brown Adhering powder and/or lumps
Titration Iodometric 97.0 to 102.0 %
Nitrate (NO3) ≤100 ppm
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤30 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤500 ppm
Zinc (Zn) ≤30 ppm
Ferrous iron (Fe 2+) Passes test (limit about 0.002%)
Phosphorous compounds (as PO4) ≤0.01 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR