Benzoesäure 99.6% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

423470020 423470250 423475000 423470100
ACRO423470020EA 249 CHF
ACRO423470020 ACRO423470250 ACRO423475000 ACRO423470100
Benzoesäure 99.6% ACS
Benzoesäure
Formel: C₇H₆O₂
Siedepunkt: 249 °C (760 mmHg)
Melting Pt: 121…123 °C
Dichte: 1,316 g/cm³ (20 °C)
Flash-Pt: 121 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002398
CAS-Nummer: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Residue after ignition ≤0.005 %
Titration with NaOH ≥99.5 %
Chlorine compounds (as Cl) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as S) ≤0.002 %
Insoluble matter ≤0.005 % (in methanol)
KMnO4-reducing substance Passes test
Freezing point 122°C to 123°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR