Valeramid 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Pentanamide

422440025 422440250 422440050
ACRO422440025EA 37.8 CHF
ACRO422440025 ACRO422440250 ACRO422440050
Valeramid 97%
Valeramid
Formel: C₅H₁₀O₂
MW: 102.13 g/mol
Melting Pt: 101…106 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00041895
CAS-Nummer: 626-97-1
EINECS: 210-974-0

Order Now

Spezifikation Testergebnisse

Appearance White Glistening powder or flakes
Infrared spectrum Conforms
Melting point 101 to 106 °C
GC ≥96.0 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR