Tetrahexylammoniumbromid 99%

Lieferant: Thermo Fisher Scientific
Achtung

420400250 420401000
ACRO420400250EA 97.3 CHF
ACRO420400250 ACRO420401000
Tetrahexylammoniumbromid 99%
Tetrahexylammoniumbromid
Formel: C₂₄H₅₂BrN
Melting Pt: 98…100 °C
Dichte: 1,0036 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011858
CAS-Nummer: 4328-13-6
EINECS: 224-363-1

Order Now

Spezifikation Testergebnisse

Appearance White to light yellow Powder or flakes
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
GC ≥98.5 %
Color scale ≤15 APHA
Stabilizer 40 to 60 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR