Sudanschwarz B, zertifiziert

Lieferant: Thermo Fisher Scientific

Synonyms: CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB

419830100
ACRO419830100EA 38.2 CHF
ACRO419830100
Sudanschwarz B, zertifiziert
Sudanschwarz B
Formel: C₂₉H₂₄N₆
MW: 456.55 g/mol
Melting Pt: 120…124 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006919
CAS-Nummer: 4197-25-5
EINECS: 224-087-1

Order Now

Spezifikation Testergebnisse

Appearance Dark brown to black Crystalline powder
Infrared spectrum Conforms
UV Lambda max 596 to 605 nm
Absorption Ratio P-15/P+15 0.98 to 1.02
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR