Stoddard solvent

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Mineral spirits

419730010 419730050
ACRO419730010EA 91.2 CHF
ACRO419730010 ACRO419730050
Stoddard solvent
Stoddard solvent
Siedepunkt: 149…213 °C (760 mmHg)
Melting Pt: –40 °C
Dichte: 0.787 g/cm³
Flash-Pt: 40 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00132767
CAS-Nummer: 8052-41-3
UN: 1300
ADR: 3,III

Order Now

Spezifikation Testergebnisse

Appearance Clear liquid
Boiling point 149 to 213°C (ASTM D 86)
Density (15.6°C) 754 to 820 kg/m³ (ASTM D 4052)
Additional info Kauri butanol value: 29 to 45 (ASTM D 1133)
Color scale ≤25 APHA (ASTM D 1209)
Bromine number ≤5 g Br/100 g (ASTM D 1159)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR