Bromkresolgrün ACS

Lieferant: Thermo Fisher Scientific

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

402930010 402930100 402930050
ACRO402930010EA 27.8 CHF
ACRO402930010 ACRO402930100 ACRO402930050
Bromkresolgrün ACS
Bromkresolgrün
Formel: C₂₁H₁₄Br₄O₅S
Melting Pt: 225 °C (dec.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005874
CAS-Nummer: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Yellow to beige to light brown Powder
Infrared spectrum Conforms
Visual transition interval from pH 3.8 (yellow) to pH 5.4 (blue)
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR