Acid orange 5 indicator grade

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: OrangeIV, Aniline yellow, 4-(4-Anilinophenylazo)benzenesulphonic acid sodium salt, CI 13080, Diphenylamine orange, Indicator, S. No.: 179, Orange IV, Tropaeolin OO

401261000 401260250 401260010
ACRO401261000EA 327 CHF
ACRO401261000 ACRO401260250 ACRO401260010
Acid orange 5 indicator grade
Acid orange 5

CAS No.: 554-73-4

Formel: C₁₈H₁₄N₃NaO₃S
MW: 375.38 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00038130
CAS-Nummer: 554-73-4

Order Now

Spezifikation Testergebnisse

Appearance Ochre to orange Powder
Infrared spectrum Conforms
Visual transition interval from pH 1.4 (red) to pH 2.6 (yellow)
GC ≥97.5 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR