6-Bromoxindol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 6-Bromo-2-oxindole, 6-Bromoindolin-2-one

399435000
ACRO399435000EA 115 CHF
ACRO399435000
6-Bromoxindol 97%
6-Bromoxindol
Formel: C₈H₆BrNO
Melting Pt: 217…221 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02179605
CAS-Nummer: 99365-40-9

Order Now

Spezifikation Testergebnisse

Appearance White to orange to brown Powder
Infrared spectrum Conforms
Melting point 215 to 220 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR