N-(tert-Butoxycarbonyl)-L-2-phenylglycine 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Boc-Phg-OH, BOC-L-alpha-phenylglycine, Boc-D-Phg-OH, N-Boc-L-2-phenylglycine

398160010 398160050
ACRO398160010EA 40.5 CHF
ACRO398160010 ACRO398160050
N-(tert-Butoxycarbonyl)-L-2-phenylglycine 99%
N-(tert-Butoxycarbonyl)-L-2-phenylglycine
Formel: C₁₃H₁₇NO₄
Melting Pt: 88…91 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00065588
CAS-Nummer: 2900-27-8

Order Now

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 88 to 91 °C
Specific optical rotation +142° to +146° (20 °C, 589 nm) (c=1%, ethanol)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR