2-Iodpyrazin 97%

Lieferant: Thermo Fisher Scientific
Achtung

397530010 397530050
ACRO397530010EA 52.6 CHF
ACRO397530010 ACRO397530050
2-Iodpyrazin 97%
2-Iodopyrazine
Formel: C₄H₃IN₂
MW: 205.99 g/mol
Siedepunkt: 109…110 °C (34 mmHg)
Flash-Pt: 110 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01319019
CAS-Nummer: 32111-21-0

Order Now

Spezifikation Testergebnisse

Appearance Colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR