n-Heptan 99+%, extra_pure

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: high purity solvents

383790025 383790050 383790010 383790250
ACRO383790025EA 148 CHF
ACRO383790025 ACRO383790050 ACRO383790010 ACRO383790250
n-Heptan 99+%, extra_pure
n-Heptan
Formel: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Siedepunkt: 98 °C (760 mmHg)
Melting Pt: –91 °C
Dichte: 0.683 g/cm³
Flash-Pt: –4 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009544
CAS-Nummer: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥99.0 %
Residue after evaporation ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR