4-Octylbenzaldehyde 97%

Lieferant: Thermo Fisher Scientific

368880050
ACRO368880050EA 160 CHF
ACRO368880050
4-Octylbenzaldehyde 97%
4-Octylbenzaldehyde

Appearance: Clear yellow Liquid

Formel: C₁₅H₂₂O
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00041872
CAS-Nummer: 49763-66-8

Order Now

Spezifikation Testergebnisse

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR