Benzothiazol-2-ylmethanol 98%

Lieferant: Thermo Scientific
Achtung

Synonyms: 2-(Hydroxymethyl)benzothiazole, 2-Benzothiazolemethanol, 2-Hydroxymethylbenzothiazole

358500010
ACRO358500010EA 49.6 CHF
ACRO358500010
Benzothiazol-2-ylmethanol 98%
Benzothiazol-2-ylmethanol
Formel: C₈H₇NOS
MW: 165,21 g/mol
Melting Pt: 95…96 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00226293
CAS-Nummer: 37859-42-0

Order Now

Spezifikation Testergebnisse

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 98 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR