Octyltriethoxysilan 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Triethoxy(octyl)silane, Triethoxyoctylsilane

338080250 338081000
ACRO338080250EA 33.5 CHF
ACRO338080250 ACRO338081000
Octyltriethoxysilan 97%
Octyltriethoxysilan
Formel: C₁₄H₃₂O₃Si
Siedepunkt: 265 °C (1013 hPa)
Melting Pt: –40 °C
Dichte: 0.88 g/cm³
Flash-Pt: 120 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00039883
CAS-Nummer: 2943-75-1
EINECS: 220-941-2

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR