(1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propandiol 96%

Lieferant: Thermo Fisher Scientific

Synonyms: L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, (1S,2S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol

334800050 334800250 334801000
ACRO334800050EA 45 CHF
ACRO334800050 ACRO334800250 ACRO334801000
(1S,2S)-2-Amino-1-(4-nitrophenyl)-1,3-propandiol 96%
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol

CAS No.: 2964-48-9

Formel: C₉H₁₂N₂O₄
Melting Pt: 160…166 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007359
CAS-Nummer: 2964-48-9

Order Now

Spezifikation Testergebnisse

Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 166 °C
Specific optical rotation +29° to +33° (20 °C, 589 nm) (c=1, 6 N HCl)
Titration with HClO4 ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR