Blei(II)acetat Trihydrat 99.999% Trace metals basis, extra_pure

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

317230250
ACRO317230250EA 183 CHF
ACRO317230250
Blei(II)acetat Trihydrat 99.999% Trace metals basis, extra_pure
Blei(II)acetat Trihydrat
Formel: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Siedepunkt: 280
Melting Pt: 75
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00150023
CAS-Nummer: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1 III
Merck Index: 13,05415
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR