2,8-Bis(trifluoromethyl)quinolin-4-ol 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 2,8-Bis(trifluoromethyl)-4-quinolinol

306010010
ACRO306010010EA 21.5 CHF
ACRO306010010
2,8-Bis(trifluoromethyl)quinolin-4-ol 99%
2,8-Bis(trifluoromethyl)quinolin-4-ol

CAS No.: 35853-41-9

Formel: C₁₁H₅F₆NO
MW: 281.16 g/mol
Melting Pt: 130…134 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00075091
CAS-Nummer: 35853-41-9

Order Now

Spezifikation Testergebnisse

Appearance White to brown-yellow to light grey Powder
Infrared spectrum Conforms
Melting point 128 to 134 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR