Metacycline hydrochloride, 98%, sec. Standard

Lieferant: Thermo Fisher Scientific

303861000
ACRO303861000EA 124 CHF
ACRO303861000
Metacycline hydrochloride, 98%, sec. Standard
Standards Organische Standards

MDL No.: MFCD04972012
Appearance: Yellow Crystalline powder
Storage Temperature: Ambient

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR