cis-4-Heptenal 96%

Lieferant: Thermo Scientific
Achtung

Synonyms: cis-4-Heptenal, Cis-4-heptenal, (Z)-4-Heptenal

298220100
ACRO298220100EA 91.2 CHF
ACRO298220100
cis-4-Heptenal 96%
cis-4-Heptenal
Formel: C₇H₁₂O
MW: 112,17 g/mol
Siedepunkt: 46 °C (13 mmHg)
Dichte: 0.849 g/cm³
Flash-Pt: 37 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00014682
CAS-Nummer: 6728-31-0
EINECS: 229-779-7
UN: 1989
ADR: 3,III

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4360 (20°C, 589 nm)
GC ≥95 %
Specific gravity (25°C) 0.843 to 0.855

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR