tri-Kaliumorthophosphat Monohydrat 96%, reinst

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Potassium phosphate monohydrate, Potassium phosphate tribasic monohydrate, Potassium phosphate hydrate

263790010 263790250 263795000 263790050 263790251
ACRO263790010EA 87.6 CHF
ACRO263790010 ACRO263790250 ACRO263795000 ACRO263790050 ACRO263790251
tri-Kaliumorthophosphat Monohydrat 96%, reinst
tri-Kaliumorthophosphat Monohydrat
Formel: K₃O₄P·1H₂O
MW: 230.28 g/mol
Melting Pt: 1340 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00150410
CAS-Nummer: 27176-10-9
EINECS: 231-907-1

Order Now

Spezifikation Testergebnisse

Appearance White Powder
Titration with HClO4 ≥95 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm
Loss on ignition ≤13 % (800°C)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR