Methyl-6-methylnicotinat 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Methyl 6-methylnicotinate

248210020 248210100
ACRO248210020EA 56.8 CHF
ACRO248210020 ACRO248210100
Methyl-6-methylnicotinat 97%
Methyl-6-methylnicotinat
Formel: C₈H₉NO₂
Siedepunkt: 108…110 °C (22.5 mmHg)
Melting Pt: 31…37 °C
Flash-Pt: 103 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006340
CAS-Nummer: 5470-70-2

Order Now

Spezifikation Testergebnisse

Appearance Brown Low melting solid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR