L(-)-Carnitin 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate

241040100 241040010 241040500
ACRO241040100EA 180 CHF
ACRO241040100 ACRO241040010 ACRO241040500
L(-)-Carnitin 99+%
L(-)-Carnitin
Formel: C₇H₁₅NO₃
MW: 161.20 g/mol
Melting Pt: 208…212 °C (Zers.)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00038747
CAS-Nummer: 541-15-1
EINECS: 208-768-0

Order Now

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
Specific optical rotation -29° to -32° (20°C, 589 nm) (c=1, H2O)
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR