Bromkresolpurpur Natriumsalz, wasserlöslich, rein Indikator

Lieferant: Thermo Fisher Scientific

Synonyms: Sodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonate, Bromocresol purple water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolate

229610050 229610100 229610250 229611000
ACRO229610050EA 31.5 CHF
ACRO229610050 ACRO229610100 ACRO229610250 ACRO229611000
Bromkresolpurpur Natriumsalz, wasserlöslich, rein Indikator
Bromkresolpurpur Natriumsalz
Formel: C₂₁H₁₅Br₂NaO₅S
MW: 562.21 g/mol
Melting Pt: 255 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00148896
CAS-Nummer: 62625-30-3
EINECS: 263-655-3

Order Now

Spezifikation Testergebnisse

Appearance Red-brown Powder
Infrared spectrum Conforms
Solubility (0.1 % in water) Clear
Specific extinction E (1%/1cm) ≥900 (590 nm, 0.01 N NaOH)
Visual transition interval from pH 5.2 (yellow) to p 6.8 (purple)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR