Pararosaniliniumchlorid, rein, hohe Reinheit biologische Färbemittel

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride

227880250 227881000 227880050 227885000
ACRO227880250EA 95.1 CHF
ACRO227880250 ACRO227881000 ACRO227880050 ACRO227885000
Pararosaniliniumchlorid, rein, hohe Reinheit biologische Färbemittel
Pararosaniliniumchlorid
Formel: C₁₉H₁₈ClN₃
Melting Pt: 268…270 °C (dec.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001657
CAS-Nummer: 569-61-9
EINECS: 209-321-2

Order Now

Spezifikation Testergebnisse

Appearance Green Crystalline powder
Infrared spectrum Conforms
Dye content ≥85 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR