Progesteron 98%

Lieferant: Thermo Fisher Scientific
Achtung

225650050 225650250 225651000
ACRO225650050EA 43.7 CHF
ACRO225650050 ACRO225650250 ACRO225651000
Progesteron 98%
Progesteron
Formel: C₂₁H₃₀O₂
Melting Pt: 126…131 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003658
CAS-Nummer: 57-83-0
EINECS: 200-350-6

Order Now

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 131 °C
Loss on drying ≤0.5 % (500 mg, 105 °C, 2 hrs)
HPLC ≥97.5 %
Specific optical rotation +186° to +194° (20 °C, 589 nm) (c=1, ethanol) (on dried substance)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR