Titan(IV)butoxid 98+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Titanium (IV) butoxide, Titanium tetrabutanolate, Tetra-n-butyl orthotitanate, Titanium (IV) n-butoxide

223190010 223190020 223192500 223195000 223190100
ACRO223190010EA 170 CHF
ACRO223190010 ACRO223190020 ACRO223192500 ACRO223195000 ACRO223190100
Titan(IV)butoxid 98+%
Titan(IV)butoxid
Formel: C₁₆H₃₆O₄Ti
Siedepunkt: 310…314 °C (760 mmHg)
Melting Pt: –75 °C
Dichte: 0.99 g/cm³
Flash-Pt: 47 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009433
CAS-Nummer: 5593-70-4
EINECS: 227-006-8
UN: 1993
ADR: 3,III

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Viscosity 50 to 90 cP (at 25°C)
Gravimetry 23.0 to 23.9 % (TiO2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR