Methylsalicylat 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Wintergreen Oil, Methyl 2-hydroxybenzoate

220490010 220490050 220495000 220490025
ACRO220490010EA 112 CHF
ACRO220490010 ACRO220490050 ACRO220495000 ACRO220490025
Methylsalicylat 99%
Methylsalicylat
Formel: 2-(HO)C₆H₄CO₂CH₃
MW: 152.15 g/mol
Siedepunkt: 222 °C (760 mmHg)
Melting Pt: 8…7 °C
Dichte: 1.1825 g/cm³
Flash-Pt: 110 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002214
CAS-Nummer: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5380 (20°C, 589 nm)
Density (20/4°C) 1.180 to 1.186 g/ml
GC ≥98.5 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR