Aluminiumchlorid Hexahydrat 99%, rein

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Aluminum chloride hexahydrate

217470010 217475000 217470025 217470100
ACRO217470010EA 143 CHF
ACRO217470010 ACRO217475000 ACRO217470025 ACRO217470100
Aluminiumchlorid Hexahydrat 99%, rein
Aluminiumchlorid Hexahydrat
Formel: AlCl₃·6H₂O
Melting Pt: ~ 100 °C
Dichte: 2.39 g/cm³
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149134
CAS-Nummer: 7784-13-6
EINECS: 231-208-1
ADR: 8,III

Order Now

Spezifikation Testergebnisse

Appearance White to yellow Adhering powder or crystals
Titration Argentometric ≥98.5 %
Water 42 to 48 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR