Cyclohexan 99.5%, reinst

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Hexahydrobenzene, Hexamethylene

210570010 210570025 210570100
ACRO210570010EA 80.7 CHF
ACRO210570010 ACRO210570025 ACRO210570100
Cyclohexan 99.5%, reinst
Cyclohexan
Formel: C₆H₁₂
Siedepunkt: 81 °C (760 mmHg)
Melting Pt: 6.5 °C
Dichte: 0.77 g/cm³
Flash-Pt: –18 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003814
CAS-Nummer: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.01 %
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR