4-Methylmorpholine N-oxide hydrate 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: NMO hydrate

208340250 208341000
ACRO208340250EA 75.8 CHF
ACRO208340250 ACRO208341000
4-Methylmorpholine N-oxide hydrate 97%
4-Methylmorpholine N-oxide hydrate
Formel: C₅H₁₁NO₂·1H₂O
MW: 117.147 g/mol (anhy)
Melting Pt: 70…75 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00005947
CAS-Nummer: 70187-32-5
EINECS: 231-391-8

Order Now

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Water ≤15 % (K.F.)
Titration with HClO4 ≥96.0 % (on dry substance)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR