Tetramethylammoniumhydroxid Pentahydrat 97%

Lieferant: Thermo Fisher Scientific
Gefahr

207520250 207521000 207525000
ACRO207520250EA 59.4 CHF
ACRO207520250 ACRO207521000 ACRO207525000
Tetramethylammoniumhydroxid Pentahydrat 97%
Tetramethylammoniumhydroxid Pentahydrat
Formel: C₄H₁₃NO·5H₂O
Melting Pt: 62…71 °C
Dichte: 1,829 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149566
CAS-Nummer: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8,II

Order Now

Spezifikation Testergebnisse

Appearance White to off-white Hygroscopic powder or crystals
Infrared spectrum Conforms
Melting point 62 to 73 °C
Titration with HCl ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR