Zirkonium(IV)chlorid, wasserfrei 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Zirconium (IV) chloride

206410050 206411000 206415000 206410025
ACRO206410050EA 50.5 CHF
ACRO206410050 ACRO206411000 ACRO206415000 ACRO206410025
Zirkonium(IV)chlorid, wasserfrei 98%
Zirkonium(IV)chlorid
Formel: Cl₄Zr
MW: 233.03 g/mol
Siedepunkt: 3577
Melting Pt: 437 °C (Subl.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011306
CAS-Nummer: 10026-11-6
EINECS: 233-058-2
UN: 2503
ADR: 8 III
Merck Index: 12,10307

Order Now

Spezifikation Testergebnisse

Appearance White Powder
Assay ≥97.5 % (ZrCl4)
Aluminium (Al) ≤100 ppm
Chromium (Cr) ≤100 ppm
Hafnium (Hf) ≤100 ppm
Iron (Fe) ≤1000 ppm
Silicon (Si) ≤150 ppm
Titanium (Ti) ≤100 ppm

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR