Ammoniaklösung 28 - 30% (w/w) in Wasser zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

205840010 205840025 205840100 205840250
ACRO205840010EA 41.2 CHF
ACRO205840010 ACRO205840025 ACRO205840100 ACRO205840250
Ammoniaklösung 28 - 30% (w/w) in Wasser zur Analyse
Ammoniaklösung
Formel: NH₃
Siedepunkt: 32…37.7 °C (1013 hPa)
Melting Pt: –57 °C
Dichte: 0.91 g/cm³
Lager Temperatur: Kühlschrank
MDL Number: MFCD00011418
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Spezifikation Testergebnisse

Appearance Clear solution
Color scale ≤10 APHA
Residue after evaporation ≤0.002 % (105 °C)
Titration with HCl 28.0 to 30.0 % (As NH3)
Chloride (Cl) ≤1 ppm
Sulfate (SO4) ≤5 ppm
Carbonate (as CO2) ≤0.006 %
Iron (Fe) ≤0.25 ppm
Heavy metals (as Pb) ≤1 ppm
Oxidizing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR