Amberlite® XAD-7 HP

Lieferant: Thermo Fisher Scientific

Synonyms: Amberlite® XAD7HP

202241000 202245000 202240025
ACRO202241000EA 40.4 CHF
ACRO202241000 ACRO202245000 ACRO202240025
Amberlite® XAD-7 HP
Amberlite® XAD-7 HP

CAS No.: 37380-43-1

Lager Temperatur: Raumtemperatur
MDL Number: MFCD00132705
CAS-Nummer: 37380-43-1

Order Now

Spezifikation Testergebnisse

Appearance White to pink-beige Beads
Particle size ≤8.0 % (> 1.180 mm)
Harmonic mean size 0.430 to 0.690 mm
Uniformity coefficient ≤2.0
TOC-AFNOR test ≤10 ppm (total)
Moisture holding capacity 61.0 to 69.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR