Hydraziniumsulfat 99+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Hydrazine sulphate salt, Hydrazine sulphate

201861000 201865000
ACRO201861000EA 48.6 CHF
ACRO201861000 ACRO201865000
Hydraziniumsulfat 99+%
Hydraziniumsulfat
Formel: NH₂NH₂·H₂SO₄
MW: 130.12 g/mol
Melting Pt: 254
MDL Number: MFCD00044873
CAS-Nummer: 10034-93-2
EINECS: 233-110-4
UN: 3288
ADR: 6.1 III
Merck Index: 14,04772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e277940036b3cf71839ff7458f0c7d08bff6f6d5&vid=7a677e6f1725cfa1b9bdef05cc04304e6739079e&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance White Crystalline powder
Titration with NaOH ≥99.0 %
Total chloride ≤20 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤5 ppm
Magnesium (Mg) ≤5 ppm
Potassium (K) ≤10 ppm
Sodium (Na) ≤20 ppm
Zinc (Zn) ≤5 ppm
Total silicon (Si) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR