Ammoniumcer(IV)nitrat 99% zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate

201441000 201442500 201440010 201440050
ACRO201441000EA 92.7 CHF
ACRO201441000 ACRO201442500 ACRO201440010 ACRO201440050
Ammoniumcer(IV)nitrat 99% zur Analyse
Ammoniumcer(IV)nitrat
Formel: (NH₄)₂Ce(NO₃)₆
Melting Pt: 107…108 °C
Dichte: 2,2 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00151121
CAS-Nummer: 16774-21-3
EINECS: 240-827-6
UN: 3085
ADR: 5.1,II
Merck Index: 13,00509

Order Now

Spezifikation Testergebnisse

Appearance Orange Crystalline powder or crystals
Solubility (10 % in water) Clear
Moisture ≤2 %
Titration ≥98.5 % (on dry substance)
Calcium oxide (CaO) ≤5 ppm
Iron(III)oxide (Fe2O3) ≤5 ppm
Potassium oxide (K2O) ≤3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR