Benzimidazol 98%

Lieferant: Thermo Fisher Scientific

Synonyms: 1H-Benzo[d]imidazole, 1H-Benzimidazole

186420250 186421000 186425000 186420025
ACRO186420250EA 27.2 CHF
ACRO186420250 ACRO186421000 ACRO186425000 ACRO186420025
Benzimidazol 98%
Benzimidazol
Formel: C₇H₆N₂
Siedepunkt: 360 °C
Melting Pt: 171…174 °C
Dichte: 1,23 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005585
CAS-Nummer: 51-17-2
EINECS: 200-081-4
Merck Index: 13,01082

Order Now

Spezifikation Testergebnisse

Appearance Beige to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 171 to 174 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR