Polydimethylsiloxan, 100 cSt

Lieferant: Thermo Fisher Scientific

Synonyms: Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone

178442500 178445000 178440025
ACRO178442500EA 129 CHF
ACRO178442500 ACRO178445000 ACRO178440025
Polydimethylsiloxan, 100 cSt
Polydimethylsiloxane
Formel: (CH₃)₃SiO(Si(CH₃)₂O)n
Siedepunkt: >65 °C (760 mmHg)
Dichte: 0.965 g/cm³
Flash-Pt: >100 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00084411
CAS-Nummer: 9016-00-6

Order Now

Spezifikation Testergebnisse

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4050 (20°C, 589 nm)
Color scale ≤35 APHA
Viscosity 95 to 105 cSt (at 25°C)
Volatiles ≤0.5 % (200°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR