1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Hexahydro-m-xylylenediamine, 1,3-Di(aminomethyl)cyclohexane, Cyclohexane-1,3-diyldimethanamine

177480050 177481000 177485000
ACRO177480050EA 42.4 CHF
ACRO177480050 ACRO177481000 ACRO177485000
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) 99%
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture)

Appearance: Clear colorless Liquid

Formel: C₈H₁₈N₂
Siedepunkt: 220 °C (760 mmHg)
Melting Pt: –70 °C
Dichte: 0.94 g/cm³
Flash-Pt: 106 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001522
CAS-Nummer: 2579-20-6
UN: 3267
ADR: 8,II

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4920 to 1.4940 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR